Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Human Recombinant NT-4 (from <i>E. coli</i>)

Human Recombinant NT-4 (from E. coli)

Human Recombinant NT-4 (from <i>E. coli</i>)
Certificates
Human Recombinant NT-4 (from <i>E. coli</i>)

4 Options Available Below

Product Details & Documents

Chemerin is a chemoattractant adipokine that is expressed in white adipose, liver, skin, and lung tissues. Chemerin is a ligand for the G protein-coupled receptor chemokine-like receptor 1 (ChemR23), which is expressed on dendritic cells, macrophages, and adipocytes.

  • High quality
  • Low endotoxin
  • Affordable

Chemerin functions to recruit macrophages to sites of tissue damage and inflammation. Chemerin is also a regulator of glucose metabolism in the liver. Due to the roles of chemerin during metabolism and inflammation, it may be a key factor in obesity-related diseases such as type two diabetes mellitus.

Delivery information: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.

Caution: For research use only.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • SHBT100-33-10UG
  • SHBT100-33-100UG
  • SHBT100-33-500UG
  • SHBT100-33-1MG
  • Protein/peptide name:
  • NT-4
  • NT-4
  • NT-4
  • NT-4
  • Protein synonyms:
  • neurotrophin 5|NT-5|NT-4/5|neurotrophic factor
  • neurotrophin 5|NT-5|NT-4/5|neurotrophic factor
  • neurotrophin 5|NT-5|NT-4/5|neurotrophic factor
  • neurotrophin 5|NT-5|NT-4/5|neurotrophic factor
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein function:
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Species:
  • Human
  • Human
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • P34130
  • P34130
  • P34130
  • P34130
  • Sequence:
  • MGVSETAPAS RRGELAVCDA VSGWVTDRRT AVDLRGREVE VLGEVPAAGG SPLRQYFFET RCKADNAEEG GPGAGGGGCR GVDRRHWVSE CKAKQSYVRA LTADAQGRVG WRWIRIDTAC VCTLLSRTGR A
  • MGVSETAPAS RRGELAVCDA VSGWVTDRRT AVDLRGREVE VLGEVPAAGG SPLRQYFFET RCKADNAEEG GPGAGGGGCR GVDRRHWVSE CKAKQSYVRA LTADAQGRVG WRWIRIDTAC VCTLLSRTGR A
  • MGVSETAPAS RRGELAVCDA VSGWVTDRRT AVDLRGREVE VLGEVPAAGG SPLRQYFFET RCKADNAEEG GPGAGGGGCR GVDRRHWVSE CKAKQSYVRA LTADAQGRVG WRWIRIDTAC VCTLLSRTGR A
  • MGVSETAPAS RRGELAVCDA VSGWVTDRRT AVDLRGREVE VLGEVPAAGG SPLRQYFFET RCKADNAEEG GPGAGGGGCR GVDRRHWVSE CKAKQSYVRA LTADAQGRVG WRWIRIDTAC VCTLLSRTGR A
  • Protease-free:
  • N
  • N
  • N
  • N
  • Animal-free:
  • N
  • N
  • N
  • N
  • Nuclease-free:
  • N
  • N
  • N
  • N
  • Carrier-free:
  • Y
  • Y
  • Y
  • Y
  • Endotoxin-free:
  • N
  • N
  • N
  • N
  • Endotoxin level:
  • Low
  • Low
  • Low
  • Low
  • Endotoxin content:
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • UniProtKB:
  • P34130
  • P34130
  • P34130
  • P34130
  • Biological activity:
  • Bioactivity assay: Proliferation of a neuroblastoma cell line stably expressing TrkB (BR6), ED50 ≤20 ng/ml, bioactivity ≥5,0×10⁴ units/mg
  • Bioactivity assay: Proliferation of a neuroblastoma cell line stably expressing TrkB (BR6), ED50 ≤20 ng/ml, bioactivity ≥5,0×10⁴ units/mg
  • Bioactivity assay: Proliferation of a neuroblastoma cell line stably expressing TrkB (BR6), ED50 ≤20 ng/ml, bioactivity ≥5,0×10⁴ units/mg
  • Bioactivity assay: Proliferation of a neuroblastoma cell line stably expressing TrkB (BR6), ED50 ≤20 ng/ml, bioactivity ≥5,0×10⁴ units/mg
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • Grade:
  • RUO
  • RUO
  • RUO
  • RUO
  • Formulation:
  • 0,1% Trifluoroacetic Acid (TFA)
  • 0,1% Trifluoroacetic Acid (TFA)
  • 0,1% Trifluoroacetic Acid (TFA)
  • 0,1% Trifluoroacetic Acid (TFA)
  • Molecular weight:
  • 14,1/28,1 kDa
  • 14,1/28,1 kDa
  • 14,1/28,1 kDa
  • 14,1/28,1 kDa
  • Reconstitution instructions:
  • Sterile water at 0,1 mg/ml
  • Sterile water at 0,1 mg/ml
  • Sterile water at 0,1 mg/ml
  • Sterile water at 0,1 mg/ml
  • Storage conditions:
  • −20 °C
  • −20 °C
  • −20 °C
  • −20 °C
  • Shipping temperature:
  • RT
  • RT
  • RT
  • RT
  • Tested applications:
  • Bioassay
  • Bioassay
  • Bioassay
  • Bioassay

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Vial
    10 µG
    Log in for availability
    Log in for availability
    1 * 10 µG
    1 * 10 µG - 4 750,00 Kč
    4 750,00 Kč
    1 * 10 µG
       4 750,00 Kč

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-33-10UG
    Protein/peptide name
    NT-4
    Protein synonyms
    neurotrophin 5|NT-5|NT-4/5|neurotrophic factor
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P34130
    Sequence
    MGVSETAPAS RRGELAVCDA VSGWVTDRRT AVDLRGREVE VLGEVPAAGG SPLRQYFFET RCKADNAEEG GPGAGGGGCR GVDRRHWVSE CKAKQSYVRA LTADAQGRVG WRWIRIDTAC VCTLLSRTGR A
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0.1
    UniProtKB
    P34130
    Biological activity
    Bioactivity assay: Proliferation of a neuroblastoma cell line stably expressing TrkB (BR6), ED50 ≤20 ng/ml, bioactivity ≥5,0×10⁴ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic Acid (TFA)
    Molecular weight
    14,1/28,1 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    −20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    100 µG
    Log in for availability
    Log in for availability
    1 * 100 µG
    1 * 100 µG - 18 350,00 Kč
    18 350,00 Kč
    1 * 100 µG
       18 350,00 Kč

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-33-100UG
    Protein/peptide name
    NT-4
    Protein synonyms
    neurotrophin 5|NT-5|NT-4/5|neurotrophic factor
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P34130
    Sequence
    MGVSETAPAS RRGELAVCDA VSGWVTDRRT AVDLRGREVE VLGEVPAAGG SPLRQYFFET RCKADNAEEG GPGAGGGGCR GVDRRHWVSE CKAKQSYVRA LTADAQGRVG WRWIRIDTAC VCTLLSRTGR A
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0.1
    UniProtKB
    P34130
    Biological activity
    Bioactivity assay: Proliferation of a neuroblastoma cell line stably expressing TrkB (BR6), ED50 ≤20 ng/ml, bioactivity ≥5,0×10⁴ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic Acid (TFA)
    Molecular weight
    14,1/28,1 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    −20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    500 µG
    Log in for availability
    Log in for availability
    1 * 500 µG
    1 * 500 µG - 91 170,00 Kč
    91 170,00 Kč
    1 * 500 µG
       91 170,00 Kč

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-33-500UG
    Protein/peptide name
    NT-4
    Protein synonyms
    neurotrophin 5|NT-5|NT-4/5|neurotrophic factor
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P34130
    Sequence
    MGVSETAPAS RRGELAVCDA VSGWVTDRRT AVDLRGREVE VLGEVPAAGG SPLRQYFFET RCKADNAEEG GPGAGGGGCR GVDRRHWVSE CKAKQSYVRA LTADAQGRVG WRWIRIDTAC VCTLLSRTGR A
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0.1
    UniProtKB
    P34130
    Biological activity
    Bioactivity assay: Proliferation of a neuroblastoma cell line stably expressing TrkB (BR6), ED50 ≤20 ng/ml, bioactivity ≥5,0×10⁴ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic Acid (TFA)
    Molecular weight
    14,1/28,1 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    −20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 151 960,00 Kč
    151 960,00 Kč
    1 * 1 mg
       151 960,00 Kč

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-33-1MG
    Protein/peptide name
    NT-4
    Protein synonyms
    neurotrophin 5|NT-5|NT-4/5|neurotrophic factor
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P34130
    Sequence
    MGVSETAPAS RRGELAVCDA VSGWVTDRRT AVDLRGREVE VLGEVPAAGG SPLRQYFFET RCKADNAEEG GPGAGGGGCR GVDRRHWVSE CKAKQSYVRA LTADAQGRVG WRWIRIDTAC VCTLLSRTGR A
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0.1
    UniProtKB
    P34130
    Biological activity
    Bioactivity assay: Proliferation of a neuroblastoma cell line stably expressing TrkB (BR6), ED50 ≤20 ng/ml, bioactivity ≥5,0×10⁴ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic Acid (TFA)
    Molecular weight
    14,1/28,1 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    −20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.