Mouse Recombinant TPO (from E. coli)
Thrombopoietin (TPO) is a growth factor that is produced by liver and kidney tissues. TPO binds the TPO receptor (CD110) to promote megakaryocyte maturation, differentiation, and the production of platelets.
- High quality
- Low endotoxin
- Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
: For research use only.
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity assay: MO7e cell proliferation, ED50 ≤5 ng/ml, bioactivity ≥2,0×10⁵ units/mg
- Gene ID:P40226
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:c-MPL ligand|MGDF|Megakaryocyte colony stimulating factor
- UniProtKB:P40226
- Protein/peptide name:TPO
- Purity:≥95%
- Molecular weight:18,8 kDa
- Sequence:MSPVAPACDP RLLNKLLRDS HLLHSRLSQC PDVDPLSIPV LLPAVDFSLG EWKTQTEQSK AQDILGAVSL LLEGVMAARG QLEPSCLSSL LGQLSGQVRL LLGALQGLLG TQLPLQGRTT AHKDPNALFL SLQQLLRGKV RFLLLVEGPT LCVRRTLPTT AVPSSTSQLL TLNKF
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Frequently Bought Together