
2-Acetyl-1H-pyrrole 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1072-83-9
:
Formula:C₆H₇NO
MDL Number:MFCD00005220
:109.13 g/mol
Boiling Pt:220 °C (760 mmHg)
Melting Pt:88…91 °C
Storage Temperature:Ambient
EINECS:214-016-2
:GHS07
Specification Test Results
- Appearance:White to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:88 to 91 °C
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together