Rat Recombinant TNFalpha (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Tumor necrosis factor alpha (TNF-α) is an inflammatory cytokine secreted by macrophages, monocytes, neutrophils, T cells, and NK-cells following stimulation by bacterial lipopolysaccharide (LPS). TNF-α signal activation occurs through two receptors, TNFR1 and TNFR2. TNFR1 is expressed on most cell types, unlike TNFR2, which is expressed mainly on immune cells. TNF-α functions to stimulate phagocytosis in macrophages, chemoattract neutrophils, increase insulin resistance, and induce fever.
- High Quality
- Low Endotoxin
- Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Rat
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity assay: Cytolysis of mouse L929 cells in the presence of Actinomycin D, ED50 ≤0,05 ng/ml, Bioactivity ≥2,0×10⁷ units/mg
- Gene ID:P16599
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:Cytotoxin|DIF|Necrosin|Cachexin|TNFSF2|TNF|Cachectin
- Protein/peptide name:TNFalpha
- Purity:≥95%
- Molecular weight:17,3 kDa
- Sequence:MLRSSSQNSS DKPVAHVVAN HQAEEQLEWL SQRANALLAN GMDLKDNQLV VPADGLYLIY SQVLFKGQGC PDYVLLTHTV SRFATSYQEK VSLLSAIKSP CPKDTPEGAE LKPWYEPMYL GGVSQLEKGD LLSAEVNLPK YLDITESGQV YFGVIAL
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together