
2,4-Dichloro-6-methylpyrimidine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:5424-21-5
:
Formula:C₅H₄Cl₂N₂
MDL Number:MFCD00006064
Boiling Pt:219 °C (1013 hPa)
Melting Pt:47 °C
EINECS:226-563-4
UN:3263
ADR:8,II
:M-Clarity™ quality level = MQ100
:GHS05
Frequently Bought Together