Order Entry
Czech Republic
ContactUsLinkComponent
4,4'-Bipyridyl, anhydrous 98%
Toxic
4,4'-Bipyridyl, anhydrous 98%
Certificates
undefined
Toxic
4,4'-Bipyridyl, anhydrous 98%

Specification Test Results

  • Appearance:
    Off-white to beige Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    110 to 114 °C
  • Water:
    ≤2.0 % (K.F.)
  • Sulfated ash:
    ≤0.1 %
  • Titration with HClO4:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together