
2,2'-Bipyridyl ACS, GR for analysis, reagent for iron(II) and molybdenum, Supelco®
Catalog # 1.03098.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Assay (perchloric acid titration):≥ 99.5 %
- Identity (IR-spectrum):passes test
- Melting point:70 - 72 °C
- Absorbance of solution (50% w/v; methanol):conforms
- Sensitivity (to ferrous ion):passes test
- Flash Point:121 °C
- pH:7.5 (5 g/l, H2O, 25 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic materials
Specification Test Results
About this item
:
:156.19 g/mol
Merck Index:13,03382
Boiling Pt:273 °C (1013 hPa)
Melting Pt:70…72 °C
Density:1.326 g/cm³ (–150 °C)
Flash Pt:121 °C
Storage Temperature:Ambient
EINECS:206-674-4
UN:2811
ADR:6.1,III
:M-Clarity™ quality level = MQ100
:GHS06
CAS Number:366-18-7
Formula:C₁₀H₈N₂
MDL Number:MFCD00006212