
2,6-Di-tert-butyl-4-methylpyridine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:38222-83-2
:
Formula:C₁₄H₂₃N
MDL Number:MFCD00006305
:205.34 g/mol
:M-Clarity™ quality level = MQ200
EINECS:253-834-4
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together