Order Entry
Czech Republic
ContactUsLinkComponent
Mouse Recombinant IL-33 (from E. coli)
Mouse Recombinant IL-33 (from E. coli)
Certificates
undefined
Mouse Recombinant IL-33 (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological activity:
    Bioactivity assay: D10S cell proliferation, ED50 ≤2 ng/ml, Bioactivity ≥5,0×10⁵ units/mg
  • Gene ID:
    Q8BVZ5
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    NP-HEV
  • Protein/peptide name:
    IL-33
  • Purity:
    ≥90%
  • Molecular weight:
    17,7 kDa
  • Sequence:
    MSIQGTSLLT QSPASLSTYN DQSVSFVLEN GCYVINVDDS GKDQEQDQVL LRYYESPCPA SQSGDGVDGK KLMVNMSPIK DTDIWLHAND KDYSVELQRG DVSPPEQAFF VLHKKSSDFV SFECKNLPGT YIGVKDNQLA LVEEKDESCN NIMFKLSKI
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate, pH 7,5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Frequently Bought Together