Order Entry
Czech Republic
ContactUsLinkComponent
2,6-Dimethoxypyridine, Sigma-Aldrich®
2,6-Dimethoxypyridine, Sigma-Aldrich®
Catalog # SIALD137006-100G
Supplier:  Merck
CAS Number:  
2,6-Dimethoxypyridine, Sigma-Aldrich®
Catalog # SIALD137006-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :139.15 g/mol
Boiling Pt:178 °C (1013 hPa)
Density:1.05 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
EINECS:228-334-4
CAS Number:6231-18-1
Formula:C₇H₉NO₂
MDL Number:MFCD00006266