
4-(Dimethylamino)pyridine for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1122-58-3
:
Formula:C₇H₁₀N₂
MDL Number:MFCD00006418
:122.17 g/mol
Boiling Pt:195 °C (1013 hPa)
Melting Pt:109…111 °C
Flash Pt:124 °C
EINECS:214-353-5
ADR:6.1,II
:M-Clarity™ quality level = MQ200
UN:2811
:GHS06
Storage Temperature:Ambient
Specification Test Results
- Solubility:60 g/l (20 °C)
- Melting Point:110 - 112 °C
- Molar Mass:122.17 g/mol
- Bulk Density:520 kg/m3
- Boiling Point:138 - 140 °C (24 hPa)
- Flash Point:110 °C
- pH:11 (60 g/l, H2O, 20 °C)
- Ignition Point:420 °C
- Hazard Class:6.1
- Hazard Class Description:Toxic materials
Specification Test Results
Related Information
Frequently Bought Together