Order Entry
Czech Republic
ContactUsLinkComponent
 
Human Recombinant IL-7 (from E. coli)
 
undefined
Human Recombinant IL-7 (from E. coli)

 

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological activity:
    Bioactivity Assay: PHA stimulated human PBMC, ED₅₀ ≤0,5 ng/ml, Bioactivity ≥2,0×10⁶ units/mg
  • Gene ID:
    P13232
  • Reconstitution instructions:
    sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    Lymphopoietin 1 (LP-1)|pre-B cell Factor
  • Protein/peptide name:
    IL-7
  • Purity:
    ≥97%
  • Molecular weight:
    17,5 kDa
  • Sequence:
    MDCDIEGKDG KQYESVLMVS IDQLLDSMKE IGSNCLNNEF NFFKRHICDA NKEGMFLFRA ARKLRQFLKM NSTGDFDLHL LKVSEGTTIL LNCTGQVKGR KPAALGEAQP TKSLEENKSL KEQKKLNDLC FLKRLLQEIK TCWNKILmgT KEH
  • Endotoxin level:
    Low
  • Formulation:
    20 mM Sodium phosphate, pH 7,5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

 

Frequently Bought Together