Human Recombinant TSLP (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Thymic stromal lymphopoietin (TSLP) is a hematopoietic cytokine produced in several tissues including the heart, liver, and prostate. TSLP induces the release of T-cell attracting chemokines from monocytes, and regulates the maturation of myeloid and epidermal dendritic cells. TSLP signals through a heterodimeric receptor complex containing the TSLP receptor (TSLP-R/CRLF2) and IL-7R alpha chain.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:Q969D9
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein/peptide name:TSLP
- Purity:≥95%
- Molecular weight:15,1 kDa
- Sequence:MYDFTNCDFE KIKAAYLSTI SKDLITYMSG TKSTEFNNTV SCSNRPHCLT EIQSLTFNPT AGCASLAKEM FAMKTKAALA IWCPGYSETQ INATQAMKKR RKRKVTTNKC LEQVSQLQGL WRRFNRPLL KQQ
- Endotoxin level:Low
- Formulation:10 mM Sodium phosphate, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together