Human Recombinant Rantes (from E. coli)
About this item
Regulated upon activation, normal T cell expressed and secreted (RANTES), also called CCL5, is a chemokine produced by T cells three to five days after T cell activation. RANTES signals through G protein-coupled receptors CCR5, CCR3, CCR1, and through the human CMV-encoded viral receptor US28. RANTES functions to recruit immune cells to inflammatory sites.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
5 Options Available Below
Product Details & Documents
Regulated upon activation, normal T cell expressed and secreted (RANTES), also called CCL5, is a chemokine produced by T cells three to five days after T cell activation. RANTES signals through G protein-coupled receptors CCR5, CCR3, CCR1, and through the human CMV-encoded viral receptor US28. RANTES functions to recruit immune cells to inflammatory sites.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- SHBT100-138-5UG
- SHBT100-138-20UG
- SHBT100-138-100UG
- SHBT100-138-500UG
- SHBT100-138-1MG
- Protein/peptide name:
- Rantes
- Rantes
- Rantes
- Rantes
- Rantes
- Protein synonyms:
- CCL5
- CCL5
- CCL5
- CCL5
- CCL5
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein function:
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Species:
- Human
- Human
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- P13501
- P13501
- P13501
- P13501
- P13501
- Sequence:
- SPYSSDTTPC CFAYIARPLP RAHIKEYFYT SGKCSNPAVV FVTRKNRQVC ANPEKKWVRE YINSLEMS
- SPYSSDTTPC CFAYIARPLP RAHIKEYFYT SGKCSNPAVV FVTRKNRQVC ANPEKKWVRE YINSLEMS
- SPYSSDTTPC CFAYIARPLP RAHIKEYFYT SGKCSNPAVV FVTRKNRQVC ANPEKKWVRE YINSLEMS
- SPYSSDTTPC CFAYIARPLP RAHIKEYFYT SGKCSNPAVV FVTRKNRQVC ANPEKKWVRE YINSLEMS
- SPYSSDTTPC CFAYIARPLP RAHIKEYFYT SGKCSNPAVV FVTRKNRQVC ANPEKKWVRE YINSLEMS
- Protease-free:
- N
- N
- N
- N
- N
- Animal-free:
- N
- N
- N
- N
- N
- Nuclease-free:
- N
- N
- N
- N
- N
- Carrier-free:
- Y
- Y
- Y
- Y
- Y
- Endotoxin-free:
- N
- N
- N
- N
- N
- Endotoxin level:
- Low
- Low
- Low
- Low
- Low
- Endotoxin content:
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:
- Bioactivity Assay: THP-1 chemotaxis, ED₅₀ ≤250 ng/ml, Bioactivity ≥4,0×10³ units/mg
- Bioactivity Assay: THP-1 chemotaxis, ED₅₀ ≤250 ng/ml, Bioactivity ≥4,0×10³ units/mg
- Bioactivity Assay: THP-1 chemotaxis, ED₅₀ ≤250 ng/ml, Bioactivity ≥4,0×10³ units/mg
- Bioactivity Assay: THP-1 chemotaxis, ED₅₀ ≤250 ng/ml, Bioactivity ≥4,0×10³ units/mg
- Bioactivity Assay: THP-1 chemotaxis, ED₅₀ ≤250 ng/ml, Bioactivity ≥4,0×10³ units/mg
- Purity:
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- Grade:
- RUO
- RUO
- RUO
- RUO
- RUO
- Formulation:
- 0,1% Trifluoroacetic acid (TFA)
- 0,1% Trifluoroacetic acid (TFA)
- 0,1% Trifluoroacetic acid (TFA)
- 0,1% Trifluoroacetic acid (TFA)
- 0,1% Trifluoroacetic acid (TFA)
- Molecular weight:
- 7,9 kDa
- 7,9 kDa
- 7,9 kDa
- 7,9 kDa
- 7,9 kDa
- Reconstitution instructions:
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Storage conditions:
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- Shipping temperature:
- RT
- RT
- RT
- RT
- RT
- Tested applications:
- Bioassay
- Bioassay
- Bioassay
- Bioassay
- Bioassay
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.SHBT100-138-5UGProtein/peptide nameRantesProtein synonymsCCL5Protein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP13501SequenceSPYSSDTTPC CFAYIARPLP RAHIKEYFYT SGKCSNPAVV FVTRKNRQVC ANPEKKWVRE YINSLEMSProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity Assay: THP-1 chemotaxis, ED₅₀ ≤250 ng/ml, Bioactivity ≥4,0×10³ units/mgPurity≥95%GradeRUOFormulation0,1% Trifluoroacetic acid (TFA)Molecular weight7,9 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT100-138-20UGProtein/peptide nameRantesProtein synonymsCCL5Protein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP13501SequenceSPYSSDTTPC CFAYIARPLP RAHIKEYFYT SGKCSNPAVV FVTRKNRQVC ANPEKKWVRE YINSLEMSProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity Assay: THP-1 chemotaxis, ED₅₀ ≤250 ng/ml, Bioactivity ≥4,0×10³ units/mgPurity≥95%GradeRUOFormulation0,1% Trifluoroacetic acid (TFA)Molecular weight7,9 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT100-138-100UGProtein/peptide nameRantesProtein synonymsCCL5Protein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP13501SequenceSPYSSDTTPC CFAYIARPLP RAHIKEYFYT SGKCSNPAVV FVTRKNRQVC ANPEKKWVRE YINSLEMSProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity Assay: THP-1 chemotaxis, ED₅₀ ≤250 ng/ml, Bioactivity ≥4,0×10³ units/mgPurity≥95%GradeRUOFormulation0,1% Trifluoroacetic acid (TFA)Molecular weight7,9 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT100-138-500UGProtein/peptide nameRantesProtein synonymsCCL5Protein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP13501SequenceSPYSSDTTPC CFAYIARPLP RAHIKEYFYT SGKCSNPAVV FVTRKNRQVC ANPEKKWVRE YINSLEMSProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity Assay: THP-1 chemotaxis, ED₅₀ ≤250 ng/ml, Bioactivity ≥4,0×10³ units/mgPurity≥95%GradeRUOFormulation0,1% Trifluoroacetic acid (TFA)Molecular weight7,9 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT100-138-1MGProtein/peptide nameRantesProtein synonymsCCL5Protein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP13501SequenceSPYSSDTTPC CFAYIARPLP RAHIKEYFYT SGKCSNPAVV FVTRKNRQVC ANPEKKWVRE YINSLEMSProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity Assay: THP-1 chemotaxis, ED₅₀ ≤250 ng/ml, Bioactivity ≥4,0×10³ units/mgPurity≥95%GradeRUOFormulation0,1% Trifluoroacetic acid (TFA)Molecular weight7,9 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay
Doporučení budou personalizována na základě vašich nákupních preferencí pouze v případě, že jste udělili souhlas aktivací možnosti "Vylepšit můj nákupní zážitek" na stránce "Osobní údaje - Nastavení ochrany osobních údajů".
V opačném případě obdržíte obecná doporučení.



