Human Recombinant Myostatin Propeptide (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Myostatin (GDF-8), a member of the TGF-beta superfamily, is a potent and negative regulator of skeletal muscle mass. The recombinant myostatin propeptide binds and inhibits myostatin to enhance the repair and regeneration of injured muscle and bone.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:O14793(24-266)
- Reconstitution instructions:Sterile 20 mM HCl at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein/peptide name:Myostatin Propeptide
- Purity:≥95%
- Molecular weight:27,8 kDa
- Sequence:MNENSEQKEN VEKEGLCNAC TWRQNTKSSR IEAIKIQILS KLRLETAPNI SKDVIRQLLP KAPPLRELID QYDVQRDDSS DGSLEDDDYH ATTETIITMP TESDFLMQVD GKPKCCFFKF SSKIQYNKVV KAQLWIYLRP VETPTTVFVQ ILRLIKPMKD GTRYTGIRSL KLDMNPGTGI WQSIDVKTVL QNWLKQPESN LGIEIKALDE NGHDLAVTFP GPGEDGLNPF LEVKVTDTPK RSRR
- Endotoxin level:Low
- Formulation:0,1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together