
Nicotinic acid (Niacin), Sigma-Aldrich®
Catalog # SIALN5410-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:123.11 g/mol
Merck Index:13,06552
Boiling Pt:258 °C (1013 hPa)
Melting Pt:142 °C
Density:1.473 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:193 °C
Storage Temperature:Ambient
EINECS:200-441-0
:GHS07
CAS Number:59-67-6
Formula:C₆H₅NO₂
MDL Number:MFCD00006391