Nicotinic acid (Niacin), Sigma-Aldrich®
SIALN0761-100G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
:
:123.11 g/mol
:13,06552
:258 °C (1013 hPa)
:142 °C
:1.473 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:193 °C
:Ambient
:200-441-0
:GHS07
:59-67-6
:C₆H₅NO₂
:MFCD00006391