Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Mouse Recombinant IL-13 (from <i>E. coli</i>)

Mouse Recombinant IL-13 (from E. coli)

Mouse Recombinant IL-13 (from <i>E. coli</i>)
Certificates
Mouse Recombinant IL-13 (from <i>E. coli</i>)

4 Options Available Below

Product Details & Documents

Interleukin 13 (IL-13) is a cytokine secreted from type 2 T helper (Th2) cells. IL-13 has overlapping functions with interleukin 4 (IL-4), including the induction of immunoglobulin E (IgE) secretion from B cells, and the inhibition of interleukin 1 beta (IL-1β), tumor necrosis factor alpha (TNF-α), interleukin 8 (IL-8), and interleukin 6 (IL-6) inflammatory cytokine expression. IL-13 also regulates immune cell inflammation in response to the pathophysiological changes of surrounding non-immune cells. The IL-13 receptor consists of the IL-4Ra and IL-13Ra1 subunits. IL-13 can also bind the IL-13Ra2 receptor with high affinity. IL-13 functions are mediated through the JAK/STAT signaling pathway. Human and mouse IL-13 are cross-reactive.

  • High Quality
  • Low Endotoxin
  • Affordable

A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  

Caution: For research use only.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • SHBT200-22-10UG
  • SHBT200-22-100UG
  • SHBT200-22-500UG
  • SHBT200-22-1MG
  • Protein/peptide name:
  • IL-13
  • IL-13
  • IL-13
  • IL-13
  • Protein synonyms:
  • NC30
  • NC30
  • NC30
  • NC30
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein function:
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Species:
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • P20109
  • P20109
  • P20109
  • P20109
  • Sequence:
  • MPVPRSVSLP LTLKELIEEL SNITQDQTPL CNGSMVWSVD LAAGGFCVAL DSLTNISNCN AIYRTQRILH GLCNRKAPTT VSSLPDTKIE VAHFITKLLS YTKQLFRHGP F
  • MPVPRSVSLP LTLKELIEEL SNITQDQTPL CNGSMVWSVD LAAGGFCVAL DSLTNISNCN AIYRTQRILH GLCNRKAPTT VSSLPDTKIE VAHFITKLLS YTKQLFRHGP F
  • MPVPRSVSLP LTLKELIEEL SNITQDQTPL CNGSMVWSVD LAAGGFCVAL DSLTNISNCN AIYRTQRILH GLCNRKAPTT VSSLPDTKIE VAHFITKLLS YTKQLFRHGP F
  • MPVPRSVSLP LTLKELIEEL SNITQDQTPL CNGSMVWSVD LAAGGFCVAL DSLTNISNCN AIYRTQRILH GLCNRKAPTT VSSLPDTKIE VAHFITKLLS YTKQLFRHGP F
  • Protease-free:
  • N
  • N
  • N
  • N
  • Animal-free:
  • N
  • N
  • N
  • N
  • Nuclease-free:
  • N
  • N
  • N
  • N
  • Carrier-free:
  • Y
  • Y
  • Y
  • Y
  • Endotoxin-free:
  • N
  • N
  • N
  • N
  • Endotoxin level:
  • Low
  • Low
  • Low
  • Low
  • Endotoxin content:
  • Endotoxin LAL (EU/µg) ≤1
  • Endotoxin LAL (EU/µg) ≤1
  • Endotoxin LAL (EU/µg) ≤1
  • Endotoxin LAL (EU/µg) ≤1
  • Biological activity:
  • Bioactivity assay: TF-1 cell proliferation, ED50 ≤10 ng/ml, Bioactivity ≥1,0×10⁵ units/mg
  • Bioactivity assay: TF-1 cell proliferation, ED50 ≤10 ng/ml, Bioactivity ≥1,0×10⁵ units/mg
  • Bioactivity assay: TF-1 cell proliferation, ED50 ≤10 ng/ml, Bioactivity ≥1,0×10⁵ units/mg
  • Bioactivity assay: TF-1 cell proliferation, ED50 ≤10 ng/ml, Bioactivity ≥1,0×10⁵ units/mg
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • Grade:
  • RUO
  • RUO
  • RUO
  • RUO
  • Formulation:
  • 0,1% Trifluoroacetic Acid (TFA)
  • 0,1% Trifluoroacetic Acid (TFA)
  • 0,1% Trifluoroacetic Acid (TFA)
  • 0,1% Trifluoroacetic Acid (TFA)
  • Molecular weight:
  • 12,3 kDa
  • 12,3 kDa
  • 12,3 kDa
  • 12,3 kDa
  • Reconstitution instructions:
  • Sterile 20 mM HCl at 0.1 mg/ml
  • Sterile 20 mM HCl at 0.1 mg/ml
  • Sterile 20 mM HCl at 0.1 mg/ml
  • Sterile 20 mM HCl at 0.1 mg/ml
  • Storage conditions:
  • –20 °C
  • –20 °C
  • –20 °C
  • –20 °C
  • Shipping temperature:
  • RT
  • RT
  • RT
  • RT
  • Tested applications:
  • Bioassay
  • Bioassay
  • Bioassay
  • Bioassay

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Vial
    10 µG
    Log in for availability
    Log in for availability
    1 * 10 µG
    1 * 10 µG - 4 750,00 Kč
    4 750,00 Kč
    1 * 10 µG
       4 750,00 Kč

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT200-22-10UG
    Protein/peptide name
    IL-13
    Protein synonyms
    NC30
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P20109
    Sequence
    MPVPRSVSLP LTLKELIEEL SNITQDQTPL CNGSMVWSVD LAAGGFCVAL DSLTNISNCN AIYRTQRILH GLCNRKAPTT VSSLPDTKIE VAHFITKLLS YTKQLFRHGP F
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤1
    Biological activity
    Bioactivity assay: TF-1 cell proliferation, ED50 ≤10 ng/ml, Bioactivity ≥1,0×10⁵ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic Acid (TFA)
    Molecular weight
    12,3 kDa
    Reconstitution instructions
    Sterile 20 mM HCl at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    100 µG
    Log in for availability
    Log in for availability
    1 * 100 µG
    1 * 100 µG - 18 350,00 Kč
    18 350,00 Kč
    1 * 100 µG
       18 350,00 Kč

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT200-22-100UG
    Protein/peptide name
    IL-13
    Protein synonyms
    NC30
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P20109
    Sequence
    MPVPRSVSLP LTLKELIEEL SNITQDQTPL CNGSMVWSVD LAAGGFCVAL DSLTNISNCN AIYRTQRILH GLCNRKAPTT VSSLPDTKIE VAHFITKLLS YTKQLFRHGP F
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤1
    Biological activity
    Bioactivity assay: TF-1 cell proliferation, ED50 ≤10 ng/ml, Bioactivity ≥1,0×10⁵ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic Acid (TFA)
    Molecular weight
    12,3 kDa
    Reconstitution instructions
    Sterile 20 mM HCl at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    500 µG
    Log in for availability
    Log in for availability
    1 * 500 µG
    1 * 500 µG - 91 170,00 Kč
    91 170,00 Kč
    1 * 500 µG
       91 170,00 Kč

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT200-22-500UG
    Protein/peptide name
    IL-13
    Protein synonyms
    NC30
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P20109
    Sequence
    MPVPRSVSLP LTLKELIEEL SNITQDQTPL CNGSMVWSVD LAAGGFCVAL DSLTNISNCN AIYRTQRILH GLCNRKAPTT VSSLPDTKIE VAHFITKLLS YTKQLFRHGP F
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤1
    Biological activity
    Bioactivity assay: TF-1 cell proliferation, ED50 ≤10 ng/ml, Bioactivity ≥1,0×10⁵ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic Acid (TFA)
    Molecular weight
    12,3 kDa
    Reconstitution instructions
    Sterile 20 mM HCl at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 151 960,00 Kč
    151 960,00 Kč
    1 * 1 mg
       151 960,00 Kč

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT200-22-1MG
    Protein/peptide name
    IL-13
    Protein synonyms
    NC30
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P20109
    Sequence
    MPVPRSVSLP LTLKELIEEL SNITQDQTPL CNGSMVWSVD LAAGGFCVAL DSLTNISNCN AIYRTQRILH GLCNRKAPTT VSSLPDTKIE VAHFITKLLS YTKQLFRHGP F
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤1
    Biological activity
    Bioactivity assay: TF-1 cell proliferation, ED50 ≤10 ng/ml, Bioactivity ≥1,0×10⁵ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic Acid (TFA)
    Molecular weight
    12,3 kDa
    Reconstitution instructions
    Sterile 20 mM HCl at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.