
2-Chloro-3-hydroxypyridine, Sigma-Aldrich®
Catalog # SIAL116203-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:129.55 g/mol
Boiling Pt:212 °C (1013 hPa)
Melting Pt:170…171 °C
:M-Clarity™ quality level = MQ100
EINECS:229-635-3
:GHS07
CAS Number:6636-78-8
Formula:C₅H₄ClNO
MDL Number:MFCD00006235