Order Entry
Czech Republic
ContactUsLinkComponent
1H-Pyrazole for synthesis, Sigma-Aldrich®
Irritant
1H-Pyrazole for synthesis, Sigma-Aldrich®
Catalog # 8.07572.0025
Supplier:  Merck
CAS Number:  
Irritant
1H-Pyrazole for synthesis, Sigma-Aldrich®
Catalog # 8.07572.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    66 - 68 °C
  • Molar Mass:
    68.07 g/mol
  • Boiling Point:
    186 - 188 °C

Specification Test Results

About this item

 :
Merck Index:12,08144
Boiling Pt:186 °C (1013 hPa)
Melting Pt:66…68 °C
Density:1.34 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
 :68.08 g/mol
Storage Temperature:Ambient
EINECS:206-017-1
 :GHS07
CAS Number:288-13-1
Formula:C₃H₄N₂
MDL Number:MFCD00005234