
1H-Pyrazole 98%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:288-13-1
:
Formula:C₃H₄N₂
MDL Number:MFCD00005234
Merck Index:12,08144
Boiling Pt:186 °C (1013 hPa)
Melting Pt:66…70 °C
Density:1,34 g/cm³ (20 °C)
:68.08 g/mol
EINECS:206-017-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to yellow Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:66 to 70 °C
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together