
2,3,5,6-Tetramethylpyrazine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1124-11-4
:
Formula:C₈H₁₂N₂
MDL Number:MFCD00006146
:136.2 g/mol
Boiling Pt:190 °C (1013 hPa)
Density:1.15 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:214-391-2
:GHS07
Storage Temperature:Ambient
Frequently Bought Together