
2,5-Dimethylpyrazine, Sigma-Aldrich®
Catalog # SIAL175420-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:108.14 g/mol
Boiling Pt:155 °C (1013 hPa)
Melting Pt:15 °C
Density:0.99 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:204-618-3
:GHS07
CAS Number:123-32-0
Formula:C₆H₈N₂
MDL Number:MFCD00006147