Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
Theobromine
Catalog # PROOMM0577.00
CAS Number:  
Irritant
Theobromine
Catalog # PROOMM0577.00
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :180.17 g/mol
Merck Index:12,09418
Melting Pt:350 °C
Density:1.419 g/cm³ (25 °C)
EINECS:201-494-2
 :GHS07
CAS Number:83-67-0
Formula:C₇H₈N₄O₂
MDL Number:MFCD00022830