Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Human Recombinant BCA-1 (from <i>E. coli</i>)

Human Recombinant BCA-1 (from E. coli)

Human Recombinant BCA-1 (from <i>E. coli</i>)
Certificates
Human Recombinant BCA-1 (from <i>E. coli</i>)

5 Options Available Below

Product Details & Documents

Galectin-1 belongs to the lectin family of carbohydrate binding proteins and binds glycans as both a monomer and a homodimer. Galectin-1 is produced in peripheral lymphoid organs and inflammatory sites. Galectin-1 plays important roles in acute and chronic inflammatory processes, cell growth, cell proliferation, and induces apoptosis of activated T cells. Galectin-1 also modulates cytokine secretion and inhibits pro-inflammatory cytokine production.

  • High quality
  • Low endotoxin
  • Affordable

Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.

Delivery information: Certificate of Analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.

Caution: For research use only.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • SHBT100-27-5UG
  • SHBT100-27-20UG
  • SHBT100-27-100UG
  • SHBT100-27-500UG
  • SHBT100-27-1MG
  • Protein/peptide name:
  • BCA-1
  • BCA-1
  • BCA-1
  • BCA-1
  • BCA-1
  • Protein synonyms:
  • BLC|CXCL13|BLR1 Ligand
  • BLC|CXCL13|BLR1 Ligand
  • BLC|CXCL13|BLR1 Ligand
  • BLC|CXCL13|BLR1 Ligand
  • BLC|CXCL13|BLR1 Ligand
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein function:
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Species:
  • Human
  • Human
  • Human
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • Q53X90
  • Q53X90
  • Q53X90
  • Q53X90
  • Q53X90
  • Sequence:
  • VLEVYYTSLR CRVQESSVFI PRRFIDRIQI LPRGNGCPRK EIIVWKKNKS IVCVDPQAEW IQRMMEVLRK RSSSTLPVPV FKRKIP
  • VLEVYYTSLR CRVQESSVFI PRRFIDRIQI LPRGNGCPRK EIIVWKKNKS IVCVDPQAEW IQRMMEVLRK RSSSTLPVPV FKRKIP
  • VLEVYYTSLR CRVQESSVFI PRRFIDRIQI LPRGNGCPRK EIIVWKKNKS IVCVDPQAEW IQRMMEVLRK RSSSTLPVPV FKRKIP
  • VLEVYYTSLR CRVQESSVFI PRRFIDRIQI LPRGNGCPRK EIIVWKKNKS IVCVDPQAEW IQRMMEVLRK RSSSTLPVPV FKRKIP
  • VLEVYYTSLR CRVQESSVFI PRRFIDRIQI LPRGNGCPRK EIIVWKKNKS IVCVDPQAEW IQRMMEVLRK RSSSTLPVPV FKRKIP
  • Protease-free:
  • N
  • N
  • N
  • N
  • N
  • Animal-free:
  • N
  • N
  • N
  • N
  • N
  • Nuclease-free:
  • N
  • N
  • N
  • N
  • N
  • Carrier-free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Endotoxin-free:
  • N
  • N
  • N
  • N
  • N
  • Endotoxin level:
  • Low
  • Low
  • Low
  • Low
  • Low
  • Endotoxin content:
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • UniProtKB:
  • Q53X90
  • Q53X90
  • Q53X90
  • Q53X90
  • Q53X90
  • Biological activity:
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • Grade:
  • RUO
  • RUO
  • RUO
  • RUO
  • RUO
  • Formulation:
  • 0,1% Trifluoroacetic Acid (TFA)
  • 0,1% Trifluoroacetic Acid (TFA)
  • 0,1% Trifluoroacetic Acid (TFA)
  • 0,1% Trifluoroacetic Acid (TFA)
  • 0,1% Trifluoroacetic Acid (TFA)
  • Molecular weight:
  • 10,2 kDa
  • 10,2 kDa
  • 10,2 kDa
  • 10,2 kDa
  • 10,2 kDa
  • Reconstitution instructions:
  • Sterile water at 0,1 mg/ml
  • Sterile water at 0,1 mg/ml
  • Sterile water at 0,1 mg/ml
  • Sterile water at 0,1 mg/ml
  • Sterile water at 0,1 mg/ml
  • Storage conditions:
  • −20 °C
  • −20 °C
  • −20 °C
  • −20 °C
  • −20 °C
  • Shipping temperature:
  • RT
  • RT
  • RT
  • RT
  • RT

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Vial
    5 µG
    Log in for availability
    Log in for availability
    1 * 5 µG
    1 * 5 µG - 1 920,00 Kč
    1 920,00 Kč
    1 * 5 µG
       1 920,00 Kč

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-27-5UG
    Protein/peptide name
    BCA-1
    Protein synonyms
    BLC|CXCL13|BLR1 Ligand
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q53X90
    Sequence
    VLEVYYTSLR CRVQESSVFI PRRFIDRIQI LPRGNGCPRK EIIVWKKNKS IVCVDPQAEW IQRMMEVLRK RSSSTLPVPV FKRKIP
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0.1
    UniProtKB
    Q53X90
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic Acid (TFA)
    Molecular weight
    10,2 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    −20 °C
    Shipping temperature
    RT
  • Vial
    20 µG
    Log in for availability
    Log in for availability
    1 * 20 µG
    1 * 20 µG - 4 750,00 Kč
    4 750,00 Kč
    1 * 20 µG
       4 750,00 Kč

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-27-20UG
    Protein/peptide name
    BCA-1
    Protein synonyms
    BLC|CXCL13|BLR1 Ligand
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q53X90
    Sequence
    VLEVYYTSLR CRVQESSVFI PRRFIDRIQI LPRGNGCPRK EIIVWKKNKS IVCVDPQAEW IQRMMEVLRK RSSSTLPVPV FKRKIP
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0.1
    UniProtKB
    Q53X90
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic Acid (TFA)
    Molecular weight
    10,2 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    −20 °C
    Shipping temperature
    RT
  • Vial
    100 µG
    Log in for availability
    Log in for availability
    1 * 100 µG
    1 * 100 µG - 11 890,00 Kč
    11 890,00 Kč
    1 * 100 µG
       11 890,00 Kč

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-27-100UG
    Protein/peptide name
    BCA-1
    Protein synonyms
    BLC|CXCL13|BLR1 Ligand
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q53X90
    Sequence
    VLEVYYTSLR CRVQESSVFI PRRFIDRIQI LPRGNGCPRK EIIVWKKNKS IVCVDPQAEW IQRMMEVLRK RSSSTLPVPV FKRKIP
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0.1
    UniProtKB
    Q53X90
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic Acid (TFA)
    Molecular weight
    10,2 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    −20 °C
    Shipping temperature
    RT
  • Vial
    500 µG
    Log in for availability
    Log in for availability
    1 * 500 µG
    1 * 500 µG - 51 310,00 Kč
    51 310,00 Kč
    1 * 500 µG
       51 310,00 Kč

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-27-500UG
    Protein/peptide name
    BCA-1
    Protein synonyms
    BLC|CXCL13|BLR1 Ligand
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q53X90
    Sequence
    VLEVYYTSLR CRVQESSVFI PRRFIDRIQI LPRGNGCPRK EIIVWKKNKS IVCVDPQAEW IQRMMEVLRK RSSSTLPVPV FKRKIP
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0.1
    UniProtKB
    Q53X90
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic Acid (TFA)
    Molecular weight
    10,2 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    −20 °C
    Shipping temperature
    RT
  • Vial
    1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 85 460,00 Kč
    85 460,00 Kč
    1 * 1 mg
       85 460,00 Kč

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-27-1MG
    Protein/peptide name
    BCA-1
    Protein synonyms
    BLC|CXCL13|BLR1 Ligand
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q53X90
    Sequence
    VLEVYYTSLR CRVQESSVFI PRRFIDRIQI LPRGNGCPRK EIIVWKKNKS IVCVDPQAEW IQRMMEVLRK RSSSTLPVPV FKRKIP
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0.1
    UniProtKB
    Q53X90
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic Acid (TFA)
    Molecular weight
    10,2 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    −20 °C
    Shipping temperature
    RT