


12-Crown-4 (1,4,7,10-tetraoxacyclododecane, lithium ionophore V) ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:294-93-9
:
Formula:C₈H₁₆O₄
MDL Number:MFCD00005103
:176.21 g/mol
Boiling Pt:68…70 °C (0.1 mmHg)
Melting Pt:17 °C
Density:1.108
Flash Pt:>109 °C (228 °F)
Storage Temperature:Ambient
EINECS:206-036-5
UN:2810
ADR:6.1,II
:GHS06
:GHS08
:GHS07
Related Information
Frequently Bought Together