Order Entry
Czech Republic
ContactUsLinkComponent
12-Crown-4 (1,4,7,10-tetraoxacyclododecane, lithium ionophore V) 97%
Toxic
12-Crown-4 (1,4,7,10-tetraoxacyclododecane, lithium ionophore V) 97%
Certificates
undefined
Toxic
12-Crown-4 (1,4,7,10-tetraoxacyclododecane, lithium ionophore V) 97%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid after melting
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4600 to 1.4630 (20°C, 589 nm)
  • GC:
    ≥96.0 %
  • Water:
    ≤5 %
  • Titration with HClO4:
    88.2 to 93.0 % (≤2.0 % (Tert.amine))

Specification Test Results

Related Information

Frequently Bought Together