Order Entry
Czech Republic
ContactUsLinkComponent
15-Crown-5 (1,4,7,10,13-pentaoxacyclopentadecane, lead ionophore V) 98%
Irritant
15-Crown-5 (1,4,7,10,13-pentaoxacyclopentadecane, lead ionophore V) 98%
Certificates
undefined
Irritant
15-Crown-5 (1,4,7,10,13-pentaoxacyclopentadecane, lead ionophore V) 98%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4640 to 1.4670 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • HPLC:
    ≥96.0 %
  • Specific optical rotation:
    +48° to +55° (20°C, 589 nm) (0.5% in Methanol)

Specification Test Results

Related Information

Frequently Bought Together