
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:17455-13-9
:
Formula:C₁₂H₂₄O₆
MDL Number:MFCD00005113
:264.32 g/mol
Merck Index:13,02630
Boiling Pt:396 °C (1013 hPa)
Melting Pt:37…39 °C
Density:1.237 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:109 °C
EINECS:241-473-5
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Melting Point:36 - 39 °C
- Molar Mass:264.32 g/mol
- Flash Point:109 °C
Specification Test Results
Frequently Bought Together