Interleukin 21 (IL-21) is a member of the common-gamma chain family of cytokines that are involved in immunoregulation. IL-21 is normally expressed by activated CD4+ T cells and is aberrantly expressed in Hodgkin lymphoma cells. The IL-21 receptor (IL-21R) activates the JAK/STAT signaling pathway and is expressed on T, B, and natural killer (NK) cells.
- High quality
- Low endotoxin
- Affordable
Within the B cell lineage, IL-21 is a switch factor regulating IgG1 and IgG3 antibody production. IL-21 also cooperates with IL-4 for the production of multiple antibody classes in B cells. IL-21 has pleiotropic effects on the proliferation, differentiation, and effector functions of B, T, NK, and dendritic cells.
Animal-Free (AF) proteins are identical to regular proteins and include additional documentation showing that they were produced with no materials of animal or human origin using ANIMAL-FREE purification processes.
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity assay: B9 cell proliferation, ED50 ≤5 ng/ml, bioactivity ≥2,0×10⁵ units/mg
- Gene ID:Q9HBE4
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- UniProtKB:Q9HBE4
- Protein/peptide name:IL-21
- Purity:≥97%
- Molecular weight:15,4 kDa
- Sequence:MQDRHMIRMR QLIDIVDQLK NYVNDLVPEF LPAPEDVETN CEWSAFSCFQ KAQLKSANTG NNERIINVSI KKLKRKPPST NAGRRQKHRL TCPSCDSYEK KPPKEFLERF KSLLQKMIHQ HLSSRTHGSE DS
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay