Mouse Recombinant MIP-3 B (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Macrophage inflammatory protein-3 beta (MIP-3 β), also called CCL19, is a chemokine that is expressed in the thymus, lymph nodes, and activated bone marrow stromal cells. MIP-3 β signals through the G protein-coupled receptor CCR7 to regulate normal lymphocyte recirculation. MIP-3 β also functions during T cell trafficking to the thymus, and in T cell and B cell homing to the lymph nodes and secondary lymphoid organs.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:Q548P0
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:CCL19|SCYA19|ELC|Exodus-3
- Protein/peptide name:MIP-3 B
- Purity:≥95%
- Molecular weight:9,2 kDa
- Sequence:GANDAEDCCL SVTQRPIPGN IVKAFRYLLN EDGCRVPAVV FTTLRGYQLC APPDQPWVDR IIRRLKKSSA KNKGNSTRRS PVS
- Endotoxin level:Low
- Formulation:0,1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together