Mouse Recombinant MIP-1 A (from E. coli)
Stromal cell-derived factor-1 alpha (SDF-1 α), also called CXCL12a, is one of two SDF-1 splice variants made by a wide variety of cells upon stimulation by inflammatory cytokines such as TNF, IL-1, and LPS. SDF-1α signals through the G protein-coupled receptor CXCR4 to recruit activated leukocytes. Human and mouse SDF-1 α share 99% sequence identity.
- High quality
- Low endotoxin
- Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
: For research use only.
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: THP-1 chemotaxis, ED50 ≤100 ng/ml, bioactivity ≥NA
- Gene ID:Q5QNW0
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:CCL3|LD78α
- UniProtKB:Q5QNW0
- Protein/peptide name:MIP-1alpha
- Purity:≥95%
- Molecular weight:7,9 kDa
- Sequence:APYGADTPTA CCFSYSRKIP RQFIVDYFET SSLCSQPGVI FLTKRNRQIC ADSKETWVQE YITDLELNA
- Endotoxin level:Low
- Formulation:0,1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Frequently Bought Together