Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
 

L(+)-Ascorbic acid, Sigma-Aldrich®

Supplier:   Merck
L(+)-Ascorbic acid, Sigma-Aldrich®
Supplier:  Merck
Certificates
L(+)-Ascorbic acid, Sigma-Aldrich®
L(+)-Ascorbic acid, Sigma-Aldrich®

5 Options Available Below

Product Details & Documents
Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Merck Index:13,00837
  • Boiling Pt:553 °C (1013 hPa)
  • Melting Pt:190…192 °C
  • Density:1.694 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:200-066-2
  • REACH:exempt
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 1 780,00 Kč
    1 780,00 Kč
    1 * 100 g
       1 780,00 Kč

  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 3 610,00 Kč
    3 610,00 Kč
    1 * 250 g
       3 610,00 Kč

  • Glass bottle for solids
    600 g
    Log in for availability
    Log in for availability
    1 * 600 g
    1 * 600 g - 8 170,00 Kč
    8 170,00 Kč
    1 * 600 g
       8 170,00 Kč

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 12 400,00 Kč
    12 400,00 Kč
    1 * 1 kg
       12 400,00 Kč

  • Glass bottle for solids
    6 kg
    Log in for availability
    Log in for availability
    1 * 6 kg
    1 * 6 kg - 63 440,00 Kč
    63 440,00 Kč
    1 * 6 kg
       63 440,00 Kč