Pig Recombinant FLT-3 Ligand (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Fms-related tyrosine kinase 3 ligand (FLT-3 ligand) is a growth factor that regulates hematopoietic cell proliferation. FLT-3 ligand signaling is transmitted through the fms-related tyrosine kinase 3 (FLT-3) receptor. FLT-3 ligand promotes the long-term expansion and differentiation of pro-B cells in the presence of interleukin 7 (IL-7) or in combination of IL-7 and interleukin 3 (IL-3).
- High Quality
- Low Endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Pig
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity assay: OCI-Aml5 cell proliferation, ED50 ≤5 ng/ml, Bioactivity ≥2,0×10⁵ units/mg
- Gene ID:D2K7D6
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:Flt3L|Fms-related tyrosine kinase 3 ligand
- Protein/peptide name:FLT-3 Ligand
- Purity:≥95%
- Molecular weight:17,3 kDa
- Sequence:MSPDCSFPHS PISSTFANTI RQLSDYLLQD YPVTVASNLQ DDELCGAFWR LVLAQRWMGQ LKTVAGSQMQ KLLEAVNTEI VFVTSCALQP LPSCLRFVQA NISHLLQDTS QQLVALKPWI TRRNFSRCLE LQCQPDPSTL LPPRSPGALE ATSLP
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together