Human Recombinant FGF-5 (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Fibroblast growth factor 5 (FGF-5) is a secreted heparin-binding growth factor that binds to FGF receptors 1 and 2 (FGFR1 and FGFR2). FGF-5 is expressed in the mesenchyme, skeletal muscles, central nervous system, and hair follicles to promote cell differentiation and proliferation. FGF-5 functions as a regulatory factor during hair elongation and skeletal muscle development.
- High Quality
- Low Endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity assay: NR6R-3T3 Proliferation with 1 ug heparin, ED50 ≤10 ng/ml, Bioactivity ≥1,0×10⁵ units/mg
- Gene ID:P12034
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:Heparin-binding growth factor 5|HBGF-5|Smag-82
- Protein/peptide name:FGF-5
- Purity:≥95%
- Molecular weight:27,7 kDa
- Sequence:MAWAHGEKRL APKGQPGPAA TDRNPIGSSS RQSSSSAMSS SSASSSPAAS LGSQGSGLEQ SSFQWSPSGR RTGSLYCRVG IGFHLQIYPD GKVNGSHEAN MLSVLEIFAV SQGIVGIRGV FSNKFLAMSK KGKLHASAKF TDDCKFRERF QENSYNTYAS AIHRTEKTGR EWYVALNKRG KAKRGCSPRV KPQHISTHFL PRFKQSEQPE LSFTVTVPEK KNPPSPIKSK IPLSAPRKNT NSVKYRLKFR FG
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate and 100 mM sodium chloride, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together