Span® 85, Span® for synthesis, Sigma-Aldrich®
Catalog # 8.40124.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 ml
- Seq:2315
Specifications
Specification Test Results
- Solubility:(25 °C) insoluble
- Molar Mass:957.51 g/mol
- Boiling Point:100 °C
- Vapor Pressure:- 1.4 hPa (20 °C)
- Flash Point:149 °C
- Density:0.95 g/cm3 (20 °C)
Specification Test Results
About this item
:
:957.51 g/mol
Boiling Pt:∼100 °C (1013 hPa)
Density:0.95 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:247-569-3
CAS Number:26266-58-0
Formula:C₆₀H₁₀₈O₈
MDL Number:MFCD00133820