Span® 60, Span® for synthesis, Sigma-Aldrich®
8.40121.0100
: Merck
:
- Pack type:Glass bottle
- Pk:100 g
- Seq:3401
- Solubility:(20 °C) insoluble
- Melting Point:53 °C
- Molar Mass:430.62 g/mol
- Boiling Point:100 °C (1013 hPa)
- Vapor Pressure:- 1.4 hPa (20 °C)
- Flash Point:149 °C
:
:430.63 g/mol
:12,08872
:∼100 °C (1013 hPa)
:53 °C
:1 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:>149 °C
:Ambient
:215-664-9
:1338-41-6
:C₂₄H₄₆O₆
:MFCD00005366