
Ruthenium(IV) oxide ≥99.95% (metals basis) Ru typ. 74%, Premion®, Electronic Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12036-10-1
:
Formula:O₂Ru
MDL Number:MFCD00074857
Melting Pt:1200 °C (subl.)
Density:6.97
Storage Temperature:Ambient
EINECS:234-840-6
UN:1479
ADR:5.1,II
:133.07 g/mol
:GHS03
Related Information
Frequently Bought Together