

Nickel (II) oxide, Technipur®
Supplier: Merck
Certificates
About this item
CAS Number:1313-99-1
:
Formula:NiO
MDL Number:MFCD00011145
:74.69 g/mol
Melting Pt:1990 °C
Density:4.6…6.67 g/cm³ (20 °C)
EINECS:215-215-7
:GHS08
:GHS07
Storage Temperature:Ambient
Frequently Bought Together