


Cobalt(II) oxide , Co >74% -325 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1307-96-6
:
Formula:CoO
MDL Number:MFCD00016031
:74.93 g/mol
Merck Index:12,02507
Melting Pt:1785 °C
Density:6,1 g/cm³ (20 °C)
EINECS:215-154-6
UN:3288
ADR:6.1,II
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Dark grey to green-brown Powder
- Additional info:May contain various amounts of Co(III)
- Purity:≥74.0 % Co
Specification Test Results
Frequently Bought Together