Cobalt(II) fluoride, ultra dry ≥99.99% (metals basis)
:10026-17-2
:
:CoF₂
:MFCD00010941
:96.93 g/mol
:14,2440
:1400 °C
:1200 °C
:4.46
:2923
:8,III
:ampouled under argon
:233-061-9
:GHS06
:GHS05
:GHS08
:GHS07
:Ambient
Frequently Bought Together