Order Entry
Czech Republic
ContactUsLinkComponent
Ruthenium(III) chloride, anhydrous 99.9% (trace metals basis)
Irritant
Corrosive
Ruthenium(III) chloride, anhydrous 99.9% (trace metals basis)
Certificates
undefined
Irritant
Corrosive
Ruthenium(III) chloride, anhydrous 99.9% (trace metals basis)

Specification Test Results

  • Appearance:
    Black Powder or crystals
  • Assay:
    48 to 50 % (Ru)
  • Total trace metal impurities:
    ≤0.1 %

Specification Test Results

Related Information

Frequently Bought Together