
Palladium(II) chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7647-10-1
:
Formula:PdCl₂
MDL Number:MFCD00003558
Merck Index:13,07058
Melting Pt:678 °C
Density:4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-596-2
UN:3260
ADR:8,III
:177.33 g/mol
:GHS07
Frequently Bought Together