Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
 
Palladium(II) chloride (59% Pd)
 
undefined
 
 
 
Palladium(II) chloride (59% Pd)
 

 

  • Appearance:
    Red-brown Powder
  • Assay:
    59 to 60 % (Pd)
  • Calcium (Ca):
    ≤30 ppm
  • Copper (Cu):
    ≤15 ppm
  • Iron (Fe):
    ≤15 ppm
  • Lead (Pb):
    ≤15 ppm
  • Magnesium (Mg):
    ≤15 ppm
  • Silicon (Si):
    ≤30 ppm
  • Silver (Ag):
    ≤30 ppm

 

 

Frequently Bought Together