Iridium(III) chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10025-83-9
:
Formula:Cl₃Ir
MDL Number:MFCD00011063
:298.58 g/mol
Merck Index:12,05101
Density:5.3 g/cm³ (20 °C)
EINECS:233-044-6
UN:3260
ADR:8,II
:M-Clarity™ quality level = MQ100
Frequently Bought Together