
Iridium(III) chloride Ir ≥ 62%, anhydrous
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10025-83-9
:
Formula:Cl₃Ir
MDL Number:MFCD00011063
:298.58 g/mol
Merck Index:14,5088
Melting Pt:763 °C (dec.)
Density:5.3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-044-6
UN:3260
ADR:8,II
:GHS05
Related Information
Frequently Bought Together