


Manganese(IV) oxide 88%, electrolytically precipitated, active
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1313-13-9
:
Formula:MnO₂
MDL Number:MFCD00003463
:86.94 g/mol
Merck Index:14,05730
Melting Pt:535 °C (dec.)
Density:5,03 g/cm³ (20 °C)
EINECS:215-202-6
UN:1479
ADR:5.1,II
:GHS03
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Black Powder
- Titration Iodometric:≥83 % (on dried substance)
- Additional info:Oxidation activity: >60% (yield)
Specification Test Results
Frequently Bought Together