Rat Recombinant Prolactin (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Prolactin is a hormone that is produced and secreted by the pituitary gland. Prolactin acts in an endocrine, paracrine, and autocrine manner. The prolactin receptor (PRLR) is expressed on many cell types, including cells of the reproductive organs, central nervous system, and breast cancer. Prolactin signal transduction occurs via JAK kinase signaling pathways. The primary function of prolactin is to regulate lactation, but prolactin also plays functional roles in the immune system and during cell growth, apoptosis, and differentiation.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Rat
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:P01237
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:PRL
- Protein/peptide name:Prolactin
- Purity:≥95%
- Molecular weight:22,7 kDa
- Sequence:MLPVCSGGDC QTPLPELFDR VVMLSHYIHT LYTDMFIEFD KQYVQDREFI AKAINDCPTS SLATPEDKEQ AQKVPPEVLLNLILSLVHSW NDPLFQLITG LGGIHEAPDA IISRAKEIEE QNKRLLEGIE KIISQAYPEA KGNEIYLVWS QLPSLQGVDE ESKDLAFYNNIRCLRRDSHK VDNYLKFLRC QIVHKNNC
- Endotoxin level:Low
- Formulation:20 mM Sodium phosphate, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together