


Chromium(III) chloride, anhydrous for synthesis, Sigma-Aldrich®
Catalog # 8.18090.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Solubility:(20 °C) slightly soluble
- Melting Point:1152 °C
- Molar Mass:158.35 g/mol
- Boiling Point:1300 °C decomposes
- Density:2.87 g/cm3 (25 °C)
Specification Test Results
About this item
:
:158.35 g/mol
Merck Index:12,02278
Melting Pt:1152 °C
Density:2.76 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-038-3
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ200
:GHS05
:GHS07
:GHS09
CAS Number:10025-73-7
Formula:Cl₃Cr
MDL Number:MFCD00010948