
Chromium(II) chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10049-05-5
:
Formula:Cl₂Cr
MDL Number:MFCD00010947
:122.9 g/mol
Merck Index:13,02264
Boiling Pt:1302 °C (1013 hPa)
Melting Pt:823…825 °C
Density:2.87 g/cm³ (20 °C)
UN:3260
ADR:8,III
:M-Clarity™ quality level = MQ200
EINECS:233-163-3
:GHS07
Storage Temperature:Ambient
Frequently Bought Together